missing translation for 'onlineSavingsMsg'
Learn More

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride 98.0+%, TCI America™

Catalog No. P08225G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08225G 5 g
P08221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08225G Supplier TCI America Supplier No. P08225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57981-02-9
Molecular Formula C7H5ClF5NO
Molecular Weight (g/mol) 249.57
MDL Number MFCD00012953
InChI Key HVMVKNXIMUCYJA-UHFFFAOYSA-N
Synonym o-2,3,4,5,6-pentafluorobenzyl hydroxylamine hydrochloride, florox reagent, o-perfluorophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-pentafluorophenyl methyl-, hydrochloride, o-pentafluorobenzyl hydroxylamine hydrochloride, o-2,3,4,5,6-pentafluorophenyl methyl hydroxylamine hydrochloride, oxy 2,3,4,5,6-pentafluorophenyl methyl amine, chloride, pfbha.hcl, acmc-1b0zv
PubChem CID 122307
IUPAC Name hydrogen O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine chloride
SMILES [H+].[Cl-].NOCC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 215°C
Color White
Quantity 5 g
Formula Weight 249.57
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride [for Oxime Preparation]