missing translation for 'onlineSavingsMsg'
Learn More

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%

Catalog No. p-7077188
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC186712500 250 mg
AC186710050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC186712500 Supplier Thermo Scientific Chemicals Supplier No. 186712500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, C7H4F5NO.HCl, CAS Number-57981-02-9 | CAS: 57981-02-9 | C7H5ClF5NO | 249.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57981-02-9
Molecular Formula C7H5ClF5NO
Molecular Weight (g/mol) 249.57
MDL Number MFCD00012953
InChI Key HVMVKNXIMUCYJA-UHFFFAOYSA-N
Synonym o-2,3,4,5,6-pentafluorobenzyl hydroxylamine hydrochloride, florox reagent, o-perfluorophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-pentafluorophenyl methyl-, hydrochloride, o-pentafluorobenzyl hydroxylamine hydrochloride, o-2,3,4,5,6-pentafluorophenyl methyl hydroxylamine hydrochloride, oxy 2,3,4,5,6-pentafluorophenyl methyl amine, chloride, pfbha.hcl, acmc-1b0zv
PubChem CID 122307
IUPAC Name O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine;hydrochloride
SMILES [H+].[Cl-].NOCC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 227.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Argentometry)
Linear Formula C6F5CH2ONH2·HCl
Quantity 250 mg
Solubility Information Solubility in water: soluble.
Formula Weight 249.57
Percent Purity 98%
Chemical Name or Material O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
EINECSNumber 261-057-7

RUO – Research Use Only