missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-(2,3,4,5,6-Pentafluorobenzyl)formaldoxime 98.0+%, TCI America™
$104.38 - $2018.78
Chemical Identifiers
| CAS | 86356-73-2 |
|---|---|
| Molecular Formula | C8H4F5NO |
| Molecular Weight (g/mol) | 225.118 |
| MDL Number | MFCD00191477 |
| InChI Key | SRTQFRQWTUMMTC-UHFFFAOYSA-N |
| PubChem CID | 602486 |
| IUPAC Name | N-[(2,3,4,5,6-pentafluorophenyl)methoxy]methanimine |
| SMILES | C=NOCC1=C(C(=C(C(=C1F)F)F)F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P1190100MG
|
TCI America
P1190100MG |
100 mg |
Each for $104.38
|
|
|||||
|
P11901G
|
TCI America
P11901G |
1 g |
Each for $626.32
|
|
|||||
|
P11905G
|
TCI America
P11905G |
5 g |
Each for $2,018.78
|
|
|||||
Chemical Identifiers
| 86356-73-2 | |
| 225.118 | |
| SRTQFRQWTUMMTC-UHFFFAOYSA-N | |
| N-[(2,3,4,5,6-pentafluorophenyl)methoxy]methanimine |
| C8H4F5NO | |
| MFCD00191477 | |
| 602486 | |
| C=NOCC1=C(C(=C(C(=C1F)F)F)F)F |
Specifications
| 86356-73-2 | |
| C8H4F5NO | |
| 100 mg | |
| C=NOCC1=C(C(=C(C(=C1F)F)F)F)F | |
| 225.118 | |
| 225.12 | |
| Crystalline Lumps |
| White | |
| MFCD00191477 | |
| SRTQFRQWTUMMTC-UHFFFAOYSA-N | |
| N-[(2,3,4,5,6-pentafluorophenyl)methoxy]methanimine | |
| 602486 | |
| ≥98.0% (GC) | |
| O-(2,3,4,5,6-Pentafluorobenzyl)formaldoxime |
Safety and Handling
TSCA : No