missing translation for 'onlineSavingsMsg'
Learn More

Nystose, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012931
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012931 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012931 Supplier Merck Emd Millipore Supplier No. 56218100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nystose is one of the smallest insulin related oligosaccharide, which can be enzymatically obtained from sucrose.

Specifications

CAS 13133-07-8
Physical Form Neat
Linear Formula C24H42O21
MDL Number MFCD00191672
Synonym beta-D-Fruf-(2->1)-beta-D-Fruf-(2->1)-beta-D-Fruf-(2<-->1)-alpha-D-Glup
Type Analytical Standard
Quantity 100 mg
Formula Weight 666.58
Percent Purity ≥98% (HPLC)
Grade Analytical Standard