missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Nystatin, USP/EP
Description
Nystatin USP / EP is a yellow to light tan powder, with characteristic odor. It is hygroscopic, and is affected by long exposure to light, heat, and air.
Specifications
Specifications
| Chemical Name or Material | Nystatin, USP/EP |
| Packaging | Jar 60ml, White, HDPE, Round, 33-400, TRIVENI-DB, CT |
| CAS | 1400-61-9 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 926.09 |
| Quantity | 50 Mega Units |
| Percent Purity | ≥ 4400 Units/mg (Nystatin) ≥ 5000 Units/mg (Nystatin) (1) |
| Grade | EP, USP |