missing translation for 'onlineSavingsMsg'
Learn More

Medisca Nystatin, USP/EP

Catalog No. 14186435
Change view
Click to view available options
Quantity:
1 billion unit
100 Mega Units
5 billion unit
500 Mega Units
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186435 1 billion unit
14185611 5 billion unit
14185281 100 Mega Units
14185393 500 Mega Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14186435 Supplier Medisca Supplier No. 024206
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nystatin USP / EP is a yellow to light tan powder, with characteristic odor. It is hygroscopic, and is affected by long exposure to light, heat, and air.

Nystatin USP / EP is a yellow to light tan powder, with characteristic odor. It is hygroscopic, and is affected by long exposure to light, heat, and air.

Specifications

Chemical Name or Material Nystatin, USP/EP
Packaging Jar 1500ml, White, HDPE, Round, 89-400, TRIVENI-DB, CT
CAS 1400-61-9
Type Drug Standard
Molecular Weight (g/mol) 926.09
Quantity 1 billion unit
Percent Purity ≥ 4400 Units/mg (Nystatin) ≥ 5000 Units/mg (Nystatin) (1)
Grade EP, USP