missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NYREN18 Monoclonal Antibody (K24024_14E2), Invitrogen™

Catalog No. MA555927
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555927 50 μL
MA556307 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555927 Supplier Invitrogen™ Supplier No. MA555927
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: TLEEQGVAHN VKAMVLELKQ SEEDARKNFQ LEEEEQNEAK LKEKQIQRTK RGLEILAKRA AETVVDPEMT PYLDIANQTG RSIRIPPSER

NYREN18 is a specific down-regulator of the NEDD8 conjugation system. It recruits NEDD8 and its conjugates to the proteasome for degradation. It also interacts with PSMD4/S5a, a member of the regulatory subunit of the 26S proteasome.

Specifications

Antigen NYREN18
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone K24024_14E2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene NUB1
Gene Accession No. Q9Y5A7
Gene Alias 4931404D21Rik; 6330412F12Rik; BS4; EGK_14356; NEDD8 ultimate buster 1; NEDD8 ultimate buster-1; negative regulator of ubiquitin like proteins 1; negative regulator of ubiquitin-like proteins 1; Nub1; NUB1L; NYREN18; NY-REN-18; NY-REN-18 antigen; Protein BS4; Renal carcinoma antigen NY-REN-18
Gene Symbols NUB1
Host Species Mouse
Immunogen Recombinant human NUB1 (immunogen range is 141-230).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51667
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less