missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NXPH3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5140152

Catalog No. PIPA5140152

Only null left

Description

Description

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GPPGSEDPER DDHEGQPRPR VPRKRGHISP KSRPMANSTL LGLLAPPGEA WGILGQPPNR PNH Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 92%.

May be signaling molecules that resemble neuropeptides. Ligand for alpha-neurexins. Target protein is a member of a family of neuropeptide-like glycoproteins.
Specifications

Chemical Identifiers

NXPH3
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human
NXPH3
KIAA1159; neurexophilin 3; neurexophilin-3; NPH3; Nxph3; UNQ687/PRO1327
NXPH3
Primary
Antigen affinity chromatography
0.05 mg/mL
Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O95157
11248
Recombinant protein corresponding to Human NXPH3. Recombinant protein control fragment (Product #RP-109369).
Antibody

Specifications

NXPH3
Polyclonal
Unconjugated
NXPH3
KIAA1159; neurexophilin 3; neurexophilin-3; NPH3; Nxph3; UNQ687/PRO1327
Rabbit
Antigen affinity chromatography
RUO
11248
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O95157
NXPH3
Recombinant protein corresponding to Human NXPH3. Recombinant protein control fragment (Product #RP-109369).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions