missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Tris-Acetate Welcome Pack and Mini Blot Modules, 3–8%

The NuPAGE Tris-Acetate Welcome Pack and Mini Blot Modules provide all of the necessary NuPAGE gels and buffers needed to begin using the Mini Gel Tank plus two Mini Blot Modules for wet transfer. Each Mini Gel Tank can accommodate up to two gels per run or up to two Mini Blot Modules for wet transfer.

Supplier:  Invitrogen™ EA0375I

 View more versions of this product

Catalog No. EA0375I


Only null left
Explore available promotions

Description

Description

The NuPAGE Tris-Acetate Welcome Pack and Mini Blot Modules provide all of the necessary NuPAGE gels and buffers needed to begin using the Mini Gel Tank plus two Mini Blot Modules for wet transfer. The Mini Gel Tank is compatible with all Invitrogen precast and handcast protein gels including Novex gels, NuPAGE gels, and Bolt Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run or up to two Mini Blot Modules for wet transfer. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use.

The NuPAGE Tris-Acetate Welcome Pack contains:
• Mini Gel Tank (Cat. No. A25977)
• NuPAGE Tris-Acetate mini gels (2 boxes, 20 gels)
• NuPAGE Tris-Acetate SDS Running Buffer, 20X (Cat. No. LA0041)
• NuPAGE LDS Sample Buffer, 4X (Cat. No. NP0007)
• NuPAGE Sample Reducing Agent, 10X (Cat. No. NP0009)
• HiMark Pre-stained Protein Standard (Cat. No. LC5699)
• 2 x Mini Blot Modules (Cat. No. B1000)

About the Mini Gel Tank
The Mini Gel Tank is compatible with Novex gels, NuPAGE gels, and Bolt Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. 

About NuPAGE Tris-Acetate gels
NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. NuPAGE Tris-Acetate gels are high performance polyacrylamide gels that simulate the denaturing or the native conditions of the traditional laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of proteins compared to traditional Tris-glycine SDS-PAGE gels.

Transfer
For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer Buffer (Cat. No. NP0006) for traditional wet transfer using the Mini Blot Module (Cat. No. B1000).

About the Mini Blot Module
The Mini Blot Module is a wet transfer device used exclusively with the Mini Gel Tank. The tank accommodates one blot module per chamber, or two blot modules total with the side-by-side layout. The Mini Blot Module is compatible all Invitrogen mini gels (blot size: 8.5 x 8 cm). The blot module accommodates one mini gel per wet transfer. The universal connection and molded gasket make the module easy to use, while the inner core of the module allows for use of less methanol-based transfer buffer per wet transfer (250 mL per gel). At the recommended conditions and constant voltage, proteins can be transferred to nitrocellulose or PVDF membranes typically in 30 to 60 minutes.

Specifications

Specifications

Mini Gel Tank
3 to 8%
Denaturing
1 kit
Mini
Tris-Acetate
15-well
40 to 500 kDa
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.