missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NUP50 Monoclonal Antibody (247CT10.2.5)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA537521

Catalog No. PIMA537521

Only null left
Explore available promotions

Description

Description

NUP50 Monoclonal Antibody for Western Blot

The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a member of the FG-repeat containing nucleoporins that functions as a soluble cofactor in importin-alpha:beta-mediated nuclear protein import. Pseudogenes of this gene are found on chromosomes 5, 6, and 14. Two transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

NUP50
Monoclonal
Conc. Not Determined
Ascites with 0.09% sodium azide; pH 7.4
Q9UKX7
NUP50
NUP50 recombinant protein.
RUO
10762
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
247CT10.2.5
Unconjugated
NUP50
1700030K07Rik; 50 kDa nucleoporin; AI413123; CTA-268H5.7; DKFZP564A043; MGC39961; Npap60; NPAP60L; nuclear pore associated protein; nuclear pore associated protein 60 kDa; nuclear pore complex protein Nup50; nuclear pore complex protein Nup50; LOW QUALITY PROTEIN: nuclear pore complex protein Nup50; nuclear pore complex protein nup50-like protein; Nuclear pore-associated protein 60 kDa-like; nuclear pore-associated protein 60L; nucleoporin 50; nucleoporin 50 kDa; Nucleoporin 50 kDa-like protein; nucleoporin 50kD; nucleoporin 50kDa; nucleoporin Nup50; nucleoprotein 50; Nucleoprotein 50kD; Nup50; NUP50 protein; OTTHUMP00000199530; PRO1146; Rtp60; Unknown (protein for MGC:134469)
Mouse
100 μL
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions