missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUP205 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150183
Description
NUP205 Polyclonal Antibody for IHC (P), Western Blot
This gene encodes a nucleoporin, which is a subunit of the nuclear pore complex that functions in active transport of proteins, RNAs and ribonucleoprotein particles between the nucleus and cytoplasm. Mutations in this gene are associated with steroid-resistant nephrotic syndrome.Chemical Identifiers
| NUP205 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q92621 | |
| 23165 | |
| Recombinant Nucleoporin 205kDa (NUP205) (Asp1735-Val2000). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| NUP205 | |
| 205 kDa nucleoporin; 3830404O05Rik; AV248391; C7orf14; KIAA0225; mKIAA0225; NPHS13; Nuclear pore complex protein Nup205; nucleoporin 205; nucleoporin 205kDa; nucleoporin Nup205; NUP205 | |
| NUP205 | |
| Primary | |
| Protein A |
Specifications
| NUP205 | |
| Polyclonal | |
| Unconjugated | |
| NUP205 | |
| 205 kDa nucleoporin; 3830404O05Rik; AV248391; C7orf14; KIAA0225; mKIAA0225; NPHS13; Nuclear pore complex protein Nup205; nucleoporin 205; nucleoporin 205kDa; nucleoporin Nup205; NUP205 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23165 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q92621 | |
| NUP205 | |
| Recombinant Nucleoporin 205kDa (NUP205) (Asp1735-Val2000). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |