missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUP155 Recombinant Rabbit Monoclonal Antibody (23GB2010)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553197
Description
NUP155 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Nucleoporins are the main components of the nuclear pore complex (NPC) of eukaryotic cells. They are involved in the bidirectional trafficking of molecules, especially mRNAs and proteins, between the nucleus and the cytoplasm. The protein does not contain the typical FG repeat sequences found in most vertebrate nucleoporins.Chemical Identifiers
| NUP155 | |
| 1.2 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75694, P37199, Q99P88 | |
| 117021, 170762, 9631 | |
| A synthesized peptide derived from human NUP155 (901-1050AA). | |
| Antibody |
| 23GB2010 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| NUP155 | |
| 155 kDa nucleoporin; ATFB15; D930027M19Rik; KIAA0791; mKIAA0791; N155; Nuclear pore complex protein Nup155; nucleoporin 155; nucleoporin 155kD; nucleoporin 155kDa; nucleoporin Nup155; NUP155; P140 | |
| NUP155 | |
| Primary | |
| Affinity chromatography |
Specifications
| NUP155 | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75694, P37199, Q99P88 | |
| NUP155 | |
| A synthesized peptide derived from human NUP155 (901-1050AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2010 | |
| Unconjugated | |
| NUP155 | |
| 155 kDa nucleoporin; ATFB15; D930027M19Rik; KIAA0791; mKIAA0791; N155; Nuclear pore complex protein Nup155; nucleoporin 155; nucleoporin 155kD; nucleoporin 155kDa; nucleoporin Nup155; NUP155; P140 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 117021, 170762, 9631 | |
| -20°C | |
| Liquid |