Learn More
Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes
Store tubes for automated picking/placing, liquid handling and other high throughput applications using Nunc Universal Latch Rack for Cryogenic Tubes.
Supplier:  Thermo Scientific™ 331827
 View more versions of this product
Catalog No. 12565703
Only null left
Description
Description
Store tubes for automated picking/placing, liquid handling and other high throughput applications using the Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes. Features a transparent cover with options for tubes of several sizes.
  • Non-sterile
Ordering Information:
Case of 10
Specifications
Chemical Identifiers
Transparent
10/pk., 10/cs.
Cryogenic Tube Storage Rack (w/Latch)
Specifications
Transparent
1 to 2 mL Cryogenic Tubes
Cryogenic Tube Storage Rack (w/Latch)
1-2mL Standard Cryotubes, non-sterile
10/pk., 10/cs.
Product Suggestions
Product Suggestions