missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Poly-D-Lysine or Collagen I Coated Multidishes

Enhance cell attachment to the culture surface with the extracellular matrix coating on Thermo Scientific™ Nunc™ Poly-D-Lysine or Collagen I Coated Multidishes.

Supplier:  Thermo Scientific™ 152025

 View more versions of this product

Catalog No. 02923200


Only null left

Description

Description

Enhance cell attachment to the culture surface with the extracellular matrix coating on Thermo Scientific™ Nunc™ Poly-D-Lysine or Collagen I Coated Multidishes, ideal for culturing fastidious cells that do not attach to the regular cell culture-treated surfaces.

  • Aseptically sterile and non-pyrogenic
  • Poly-D-Lysine is a chemically synthesized animal-free material
  • Collagen I is of animal origin
Warranty: 90 days
Specifications

Chemical Identifiers

Multi Dish
20
128 mm
5
Aseptically Manufactured
86 mm

Specifications

1.9 sq. cm
1 mL
86 mm
20
Poly-D-Lysine
24
86 mm
1 mL (Well)
Multi Dish
5
Clear
Aseptically Manufactured
128 mm
Round, Flat Bottom (F-well)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.