Learn More
Thermo Scientific™ Nunc™ PETG Roller Bottles
Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor.
Supplier:  Thermo Scientific™ 216005
 View more versions of this product
Catalog No. 12565530
Only null left
Description
Description
Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor with Thermo Scientific™ Nunc™ PETG Roller Bottles. Virtually unbreakable PETG provides added assurance that valuable contents are safe during laboratory and industrial-scale applications including vaccines, cell culture expansion and production of other biologics.
Key Features:
  • Molded of durable PETG
  • Can be frozen down to -40°C — supports freeze thaw cell release methods, reducing reliance on trypsin
  • Provides solutions for all scales with wide range of surface areas from 1050cm2 to 4200cm2
  • Reduces wrist strain and increases productivity with quick-action ergonomic closure
  • Can be traced by lot number on every bottle
Recommended for:
  • Cell culture expansion
  • Vaccine production
  • Antibody production
Specifications
Chemical Identifiers
Clear
Yes
Round
Quick-Action Ergonomic
PETG
Yes
Roller Bottle
Specifications
Pleated
2160-05
20
5/sleeve, 20/case
Yes
Round
Quick-Action Ergonomic
Clear
PETG
5
2100 cm2
Yes
Roller Bottle
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.