missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Multidishes with UpCell™ Surface

Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins.

$1,235.23 - $1,900.07

Specifications

Type Multi Dish
No. per Pack 1
No. per Case 6
Color Clear
Sterility Sterile
View More Specs

Products 4
Catalog Number Mfr. No. No. of Wells Culture Area Price Quantity & Availability  
Catalog Number Mfr. No. No. of Wells Culture Area Price Quantity & Availability  
03150019
View Documents Promotion Details
Thermo Scientific™
174898
48 1.1 sq. cm
Case of 6 Each for $1,723.14
Only null left
 
03150020
View Documents Promotion Details
Thermo Scientific™
174899
24 1.9 sq. cm
Case of 6 Each for $1,235.23
Only null left
 
03150021
View Documents Promotion Details
Thermo Scientific™
174900
12 3.5 sq. cm
Case of 6 Each for $1,882.75
Only null left
 
03150022
View Documents Promotion Details
Thermo Scientific™
174901
6 9.6 sq. cm
Case of 6 Each for $1,900.07
Only null left
 
Description

Description

Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with the temperature-responsive surface of Thermo Scientific™ Nunc™ UpCell™ Dishes. Simply transfer the culture to room temperature to collect cells in suspension or in the form of cell sheet with the help of a supportive membrane.

Features:
  • The hydrophicility of the UpCell surface changes with the external temperature, allowing cells to detach
  • No need for dissociation enzymes, ensuring intact cell surface proteins
  • No need for physical scraping, resulting in high cell viability
  • Supportive membranes included in the 6-well plates allow for harvesting of the cell sheets
  • Customized bar coding available
  • Shelf Life: 3 years from date of manufacturing

Recommended for:

  • Culture passaging
  • Subsequent single-cell analyses (e.g. flow cytometry)
  • 3D tissue modeling for transplantation
  • For research use only
Warranty: 90 days

 

Specifications

Specifications

Multi Dish
6
Sterile
1
Clear
Rectangular, Flat Bottom (F-well)
Videos
SDS
Documents

Documents

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. For research and single-use only