missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ MicroWell™ MiniTrays
Create more space in lab freezers and inubators with Nunc MicroWell MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
$954.55 - $5784.25
Specifications
| Bottom Shape | Flat |
|---|---|
| Material | Polystyrene |
| Shape | Rectangular |
| Type | Cell Culture Dish |
| Length (Metric) | 84 mm |
| Catalog Number | Mfr. No. | Well Count | Closure Included | Sterility | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Well Count | Closure Included | Sterility | Price | Quantity | |||
|
12-565-154
|
Thermo Scientific™
438733 |
72 | Yes | Non-sterile |
Case of 100 Each for $954.55
|
|
|||
|
12-565-155
|
Thermo Scientific™
439225 |
60 | Yes | Non-sterile |
Case of 100 Each for $1,225.28
|
|
|||
|
12-565-158
|
Thermo Scientific™
452256 |
60 | Yes | Sterile |
Case of 480 Each for $5,784.25
|
|
|||
|
12-565-157
|
Thermo Scientific™
470378 |
72 | Yes | Non-sterile |
Case of 480 Each for $5,737.31
|
|
|||
|
12-565-159
|
Thermo Scientific™
472400 |
60 | No | Sterile |
Case of 400 Each for $4,147.91
|
|
|||
Description
Create more space in lab freezers and inubators with Thermo Scientific™ Nunc™ MicroWell™ MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
- Limited adsorption of proteins
- Lids have keyed tabs for proper orientation
- Large raised alphanumeric coordinates
- Unique surface treatment to maximize sample and reagent mixing
- MiniTrays for serological applications have slightly smaller well dimensions
Specifications
| Flat | |
| Rectangular | |
| 84 mm | |
| 59 mm |
| Polystyrene | |
| Cell Culture Dish | |
| 59 mm |