Learn More
Thermo Scientific™ Nunc™ Microplate Plastic Storage Racks
Conveniently and safely store and transport samples in microplate, deep-well plates and cryobank racks by using Thermo Scientific™ Nunc™ Plastic Storage Racks.
$793.67 - $2,817.50
Specifications
Color Green
Material Polypropylene, Metal
Shape Rectangular
Product Type Microplate Rack
View More Specs
Products 3
Catalog Number Mfr. No. Array Price Quantity & Availability  
Catalog Number Mfr. No. Array Price Quantity & Availability  
12565250
View Documents Promotion Details
Thermo Scientific™
344260
10 Tall
Each for $793.67
Only null left
12566158
View Documents Promotion Details
Thermo Scientific™
344280
10 x 5
Each for $2,798.32
Only null left
12566174
View Documents Promotion Details
Thermo Scientific™
344380
5 x 5
Each for $2,817.50
Only null left
Description
Description
Conveniently and safely store and transport samples in microplate, deep-well plates and cryobank racks by using Thermo Scientific™ Nunc™ Plastic Storage Racks, ideal for genome library storage, DNA and RNA libraries, and storage of compounds for high throughput screening.
  • Horizontal design fits incubator and chest and upright freezers.
  • Retainer clip holds plates securely in place.
  • Handles on top and side offer easy transport.
  • Locking bar keeps plates secure.
  • Racks are freezer compatible down to -80°C.
  • Racks are not for use in liquid/vapor nitrogen.
Warranty:
90 days
Specifications
Specifications
Green
Polypropylene, Metal
Rectangular
Microplate Rack
Videos
SDS
Documents
Documents
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.