missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ IVF Dishes
Description
Carry out IVF applications using high-quality, reliable Thermo Scientific™ Nunc™ IVF Dishes.
Product features for reliable results:
- Facilitate gas exchange with vented lid
- Available in 35mm, 60mm and 90mm sizes
- Certificates of conformity available for every lot
Rigorously tested, certified and validated for IVF:
- CE-marked (Notified Body 2460), FDA-listed
- Produced under ISO 13485 with full lot traceability
- Sterile (SAL 10-6)
- Non-pyrogenic (LAL tested)
- USP Class VI polystyrene
- Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)
Warranty: 90 days
Specifications
Specifications
| Certifications/Compliance | CE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested |
| Description | IVF Dish, 35mm, Lid w/Airvent |
| Culture Area | 8.8 cm2 |
| Material | Polystyrene (USP Class VI) |
| Sterility | Sterile |
| Surface Treatment | Untreated |
| No. per Case | 500 |
| No. per Pack | 10 |
| Height (Metric) | 11 mm |
| Diameter (Metric) | 35 mm |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.