Learn More
Thermo Scientific™ Nunc™ EZFlip™ Conical Centrifuge Tubes
Thermo Scientific™ Nunc™ EZFlip™ Conical Centrifuge Tubes can be opened and closed with one hand due to a proprietary hinged-cap system. The tubes feature an ergonomic design, and are compatible with both fixed-angle and swing bucket centrifuge rotors.
$1,249.96 - $1,565.07
Specifications
Material Polypropylene
No. per Case 500
Shape Conical
Bottom Shape Conical
Graduated Yes
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Sterility Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Sterility Packaging Price Quantity & Availability  
12565801
View Documents Promotion Details
Thermo Scientific™
362694
15 mL Sterile Bulk
Case of 500 Each for $1,402.07
Only null left
02923219
View Documents Promotion Details
Thermo Scientific™
362694NS
15 mL Non-sterile Bulk
Case of 500 Each for $1,249.96
Only null left
12565802
View Documents Promotion Details
Thermo Scientific™
N362695
15 mL Sterile Racked
Case of 500 Each for $1,409.25
Only null left
02923220
View Documents Promotion Details
Thermo Scientific™
362695NS
15 mL Non-sterile Racked
Case of 500 Each for $1,395.39
Only null left
02923221
View Documents Promotion Details
Thermo Scientific™
362696NS
50 mL Non-sterile Bulk
Case of 500 Each for $1,357.36
Only null left
02923222
View Documents Promotion Details
Thermo Scientific™
362697NS
50 mL Non-sterile Racked
Case of 500 Each for $1,565.07
Only null left
Description
Description
Thermo Scientific™ Nunc™ EZFlip™ Conical Centrifuge Tubes can be opened and closed with one hand due to a proprietary hinged-cap system. The tubes feature an ergonomic design, and are compatible with both fixed-angle and swing bucket centrifuge rotors.
15-mL tube features
  • Integrated leakproof cap prevents cross-contamination and lost closures
  • Clarified polypropylene combines chemical resistance and high-speed tolerances
  • Writing area and flat caps for sample identification
  • Volume markings and graduations
  • DNase/RNase free
  • Product raw materials are biocompatible per USP Class VI test
  • Product is non-cytotoxic and non-pyrogenic
  • Sterilized version available (SAL 10-6)
  • Max RCF: 8500 x g for the fixed-angle rotor, 7000 x g for swing bucket rotor*
  • Operating temperature: –20 to 37°C
  • Shelf life: 2 years
50-mL tube features
  • Integrated leakproof cap prevents cross-contamination and lost closures
  • Clarified polypropylene combines chemical resistance and high-speed tolerances
  • Writing area and flat caps for sample identification
  • Volume markings and graduations
  • Product raw materials are biocompatible per USP Class VI test
  • Product is non-cytotoxic and non-pyrogenic
  • Max RCF: 15000 x g*
  • Operating temperature: –20 to 37°C
*When fully supported by conical rotor cavity or conical adaptor
Ordering Information
Tubes are supplied in a bulk pack (sleeve) or a rack
Specifications
Specifications
Polypropylene
Conical
Yes
500
Conical
Centrifuge Tube