Learn More
Thermo Scientific™ Nunc™ Disposable Loops and Needles
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
$817.64 - $3,786.11
| Catalog Number | Mfr. No. | Volume | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume | Quantity | Price | Quantity & Availability | ||||
|
130754A
|
Thermo Scientific™
251586 |
10 μL | Carton of 1000 |
Pack of 1000 Each for $999.25
Case of 4 Pack for $3,997.00
|
|
||||
|
130754B
|
Thermo Scientific™
253287 |
1 μL | Carton of 1000 |
Case of 1000 Each for $3,786.11
|
|
||||
|
13079C
|
Thermo Scientific™
254410 |
1 μL | Carton of 600 |
Pack of 600 Each for $836.46
Case of 4 Pack for $3,345.84
|
|
||||
|
130754C
|
Thermo Scientific™
254437 |
10 μL | Carton of 600 |
Pack of 600 Each for $817.64
Case of 4 Pack for $3,270.56
|
|
||||
Description
Easily identify color-coded Thermo Scientific™ Nunc™ Disposable Loops and Needles, precision molded from nontoxic (USP Class VI) polystyrene material to provide a smooth loop surface. Thermo Scientific Nunc products are high quality plastics for use in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits.
Features:
- Radiation sterilized—no need for flame sterilization
- Hydrophilic treatment during manufacturing process
- Liquid adherence properties that prevent accidental discharge of droplets
- Loops available in 10 and 1 μL sizes
- Meet semi-quantitative standard for handling samples
- Calibration documents available for any batch upon request
Specifications
| Polystyrene | |
| Sterile | |
| Single-use |