Learn More
Thermo Scientific™ Nunc™ Disposable Loops and Needles
Catalog No. 13079C
Change view
Click to view available options
Volume:
1 μL
10 μL
Quantity:
Carton of 1000
Carton of 600
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume Quantity
13079C 1 μL Carton of 600
130754A 10 μL Carton of 1000
130754B 1 μL Carton of 1000
130754C 10 μL Carton of 600
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13079C Supplier Thermo Scientific™ Supplier No. 254410
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Easily identify color-coded Thermo Scientific™ Nunc™ Disposable Loops and Needles, precision molded from nontoxic (USP Class VI) polystyrene material to provide a smooth loop surface. Thermo Scientific Nunc products are high quality plastics for use in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits.
Features:
  • Radiation sterilized—no need for flame sterilization
  • Hydrophilic treatment during manufacturing process
  • Liquid adherence properties that prevent accidental discharge of droplets
  • Loops available in 10 and 1 μL sizes
  • Meet semi-quantitative standard for handling samples
  • Calibration documents available for any batch upon request
Specifications
Disposable Single-use
Volume 1 μL
Material Polystyrene
Sterility Sterile
Color Clear
Product Type Inoculating Loop (Clear)
Quantity Carton of 600