missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ IVF Center Well Dish

Catalog No. 12565024
Change view
Click to view available options
Diameter (Metric):
55 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
12565024 55 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12565024 Supplier Thermo Scientific™ Supplier No. 150260
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Experience consistently reliable, high-quality results during embryo and oocyte washing with Thermo Scientific™ Nunc Center Well Dishes.

Thermo Scientific™ Nunc Center Well Dishes provide consistently reliable, IVF-quality results during embryo and oocyte washing.

Innovative features for quality and ease-of-use:

  • Perfect angle of reservoir well walls for locating and easy washing of samples
  • True flat bottom for maintaining optimal temperature
  • Easy grip design for increased safety

Rigorously tested, certified and validated for IVF:

  • CE-marked (Notified Body 2797), FDA-listed
  • Produced under ISO 13485 with full lot traceability

Certificates of conformity for your assurance on every lot:

  • Sterile (SAL 10-6)
  • Non-pyrogenic (LAL test)
  • USP Class VI test
  • Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA)

Warranty: 90 days

Specifications

Certifications/Compliance CE-marked, FDA-cleared, MEA-tested
Description Center Well Dish, Lid w/Airvent
Bottom Shape Flat
Color Clear
Material Polystyrene (USP Class VI)
Sterility Sterile
Surface Treatment Untreated
Type Dish
Well Count 1
No. per Case 120
No. per Pack 1
Height (Metric) 16 mm
Diameter (Metric) 55 mm
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.