Learn More
Thermo Scientific™ Cryo Vial Closure Color Coders
GREENER_CHOICE
Make it easier to identify specific vials quickly and simplify your inventory by using these color coders that fit into the top of cryogenic vial or tube closures.
$72.99 - $780.35
Specifications
Product Type Vial Closure
View More Specs
Products 16
Catalog Number Mfr. No. Color Product Line Price Quantity & Availability  
Catalog Number Mfr. No. Color Product Line Price Quantity & Availability  
12565242
View Documents Promotion Details
Thermo Scientific™
354755
White Nunc
Pack of 500 Each for $195.09
Case of 4 Pack for $780.36
Only null left
12565243
View Documents Promotion Details
Thermo Scientific™
N354879
Blue Nunc
Pack of 500 Each for $175.47
Case of 4 Pack for $701.88
Only null left
12565244
View Documents Promotion Details
Thermo Scientific™
354968
Red Nunc
Pack of 500 Each for $198.10
Case of 4 Pack for $792.40
Only null left
12565245
View Documents Promotion Details
Thermo Scientific™
355018
Green Nunc
Pack of 500 Each for $198.10
Case of 4 Pack for $792.40
Only null left
12565246
View Documents Promotion Details
Thermo Scientific™
355077
Yellow Nunc
Pack of 500 Each for $198.10
Case of 4 Pack for $792.40
Only null left
12565247
View Documents Promotion Details
Thermo Scientific™
355158
Orange Nunc
Pack of 500 Each for $198.10
Case of 4 Pack for $792.40
Only null left
12565251
View Documents Promotion Details
Thermo Scientific™
375868
Brown Nunc
Pack of 500 Each for $204.15
Case of 4 Pack for $816.60
Only null left
12565252
View Documents Promotion Details
Thermo Scientific™
375884
Pink Nunc
Pack of 500 Each for $195.09
Case of 4 Pack for $780.36
Only null left
12565253
View Documents Promotion Details
Thermo Scientific™
375906
Gray Nunc
Case of 2000 Each for $780.35
Only null left
12565254
View Documents Promotion Details
Thermo Scientific™
375922
Purple Nunc
Case of 2000 Each for $780.35
Only null left
12565180
View Documents Promotion Details
Thermo Scientific™
375930
Assorted Nunc
Pack of 500 Each for $249.90
Case of 4 Pack for $999.60
Only null left
1535047
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
50450003
Blue Nalgene
Pack of 100 Each for $72.99
Case of 5 Pack for $364.95
Only null left
1535045
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
50450000
White Nalgene
Pack of 100 Each for $72.99
Case of 5 Pack for $364.95
Only null left
1535046
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
50450002
Yellow Nalgene
Pack of 100 Each for $73.11
Case of 5 Pack for $365.55
Only null left
1535048
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
50450004
Green Nalgene
Pack of 100 Each for $72.99
Case of 5 Pack for $364.95
Only null left
1535049
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
50450005
Red Nalgene
Pack of 100 Each for $96.27
Case of 5 Pack for $481.35
Only null left
Description
Description
Make it possible to identify specific vials quickly and simplify your inventory using color coders that fit into the top of Nalgene™ Cryogenic vial closures or Nunc™ Cryogenic Tube closures.
  • Nalgene™ color coders are made from polystyrene and fit only in Nalgene™ cryogenic vial closures. Available in 5 colors
  • Nunc™ color coders are made from polycarbonate and fit only in Nunc™ cryogenic tube closures and Cryobank 2ml and 5mL tube caps. Available in 10 colors or as an assortment.
Specifications
Specifications
Vial Closure