missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cryo Vial Closure Color Coders

Catalog No. 12565242
Change view
Click to view available options
Color:
Assorted
Blue
Brown
Gray
Green
Orange
Pink
Purple
Red
White
Yellow
Product Line:
Nalgene
Nunc
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Line
12565242 White Nunc
12565243 Blue Nunc
12565244 Red Nunc
12565245 Green Nunc
12565246 Yellow Nunc
12565247 Orange Nunc
12565251 Brown Nunc
12565252 Pink Nunc
12565253 Gray Nunc
12565254 Purple Nunc
12565180 Assorted Nunc
1535047 Blue Nalgene
1535045 White Nalgene
1535046 Yellow Nalgene
1535048 Green Nalgene
1535049 Red Nalgene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 12565242 Supplier Thermo Scientific™ Supplier No. 354755

Make it easier to identify specific vials quickly and simplify your inventory by using these color coders that fit into the top of cryogenic vial or tube closures.

Make it possible to identify specific vials quickly and simplify your inventory using color coders that fit into the top of Nalgene™ Cryogenic vial closures or Nunc™ Cryogenic Tube closures.

  • Nalgene™ color coders are made from polystyrene and fit only in Nalgene™ cryogenic vial closures. Available in 5 colors
  • Nunc™ color coders are made from polycarbonate and fit only in Nunc™ cryogenic tube closures and Cryobank 2ml and 5mL tube caps. Available in 10 colors or as an assortment.

Specifications

Description Nunc™ CryoColor Vial Closure Coder, PS
Color White
Material Polystyrene
Product Line Nunc
Quantity 500/Pk., 2000/Cs.
For Use With (Equipment) Cryogenic Vial
Product Type Vial Closure
Are your Nunc CryoColor Vial Closure Coders removable?

Once inserted, the Nunc CryoColor Vial Closure Coders cannot be removed.