missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ IVF Multi Dish, 4-well
Description
Generate consistently reliable results during IVF and embryo vitrification with Thermo Scientific™ Nunc™ 4-Well IVF Dishes, which meet the rigorous requirements of IVF testing, certification and validation.
Innovative features for high quality and ease-of-use:
- Ensures proper well orientation and prevents cross contamination with notched lid and dish
- Prevents scratches on well bottoms and promotes fast temperature equilibrium with 0.8mm raised edge
- The Nunclon Delta surface treatment ensures consistency of a more hydrophilic surface
- The non-treated hydrophobic surface facilitates microdrop embryo culture
- Certificates of conformity available for every lot
Rigorously tested, certified and validated for IVF:
- CE-marked (Notified Body 2460), FDA-listed
- Produced under ISO 13485 with full lot traceability
- Sterile (SAL 10-6)
- Non-pyrogenic (LAL tested)
- USP Class VI polystyrene
- Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)
Warranty: 90 days
Specifications
Specifications
| Certifications/Compliance | CE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested |
| Description | Nunclon Delta-treated IVF Dish, 4-well |
| Culture Area | 1.9 cm2/well |
| Material | Polystyrene (USP Class VI) |
| Sterility | Sterile |
| Surface Treatment | Nunclon Delta |
| Well Count | 4 |
| No. per Case | 120 |
| No. per Pack | 4 |
| Length (Metric) | 66 mm |
| Show More |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.