missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nudel Monoclonal Antibody (K52041_9G9), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA556001
Description
Sequence of this protein is as follows: SLKEETAYWK ELSLKYKQSF QEARDELVEF QEGSRELEAE LEAQLVQAEQ RNRDLQADNQ RLKYEVEALK EKLEHQYAQS YKQVSVLEDD LSQTRAIKEQ LHKYVRELEQ ANDDLERAKR ATIVSLEDFE QRLNQAIERN AFLESELDEK ESLLVSVQRL KDEARDLRQE LAVRERQQEV TRKSAPSSPT LDCEKMDSAV QASLSLPATP
This gene encodes a thiol-activated peptidase that is phosphorylated in M phase of the cell cycle. Phosphorylation regulates the cell cycle-dependent distribution of this protein, with a fraction of the protein bound strongly to centrosomes in interphase and localized to mitotic spindles in early M phase. Overall, this protein plays a role in nervous system development. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
| Nudel | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q78PB6, Q9ERR1, Q9GZM8 | |
| Ndel1 | |
| Recombinant human NDEL1 (immunogen range is 11-220). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| K52041_9G9 | |
| Unconjugated | |
| Ndel1 | |
| 2600006O07Rik; ATP-sensitive inward rectifier potassium channel 15; DKFZp451M0318; endooligopeptidase A; EOPA; Inward rectifier K(+) channel Kir4.2; Kcnj15; Kir4.2; LIS1-interacting protein NUDEL; LIS1-interacting protein NUDEL; endooligopeptidase A; MITAP1; Mitosin-associated protein 1; mitosin-associated protein MITAP1; mNudel; mNudE-L; NDE1L1; NDE2; Ndel1; nuclear disribution gene E like; nuclear distribution gene E-like homolog 1; nuclear distribution gene E-like homolog 1 (A. nidulans); nuclear distribution protein nudE-like 1; nudE neurodevelopment protein 1 like 1; nudE neurodevelopment protein 1-like 1; nudE nuclear distribution E homolog-like 1; nudE nuclear distribution gene E homolog like 1; nudE nuclear distribution gene E homolog-like 1; Nude2; NUDEL; NUDE-like protein; Potassium channel, inwardly rectifying subfamily J member 15; potassium channel, inwardly rectifying subfamily J, member 15; potassium inwardly-rectifying channel, subfamily J, member 15; Protein mNudE-like; Protein Nudel; unnamed protein product | |
| Mouse | |
| Protein A | |
| RUO | |
| 170845, 81565, 83431 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |