missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nuclear Receptor Buffer F
Description
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
Specifications
Specifications
| Chemical Name or Material | Nuclear Receptor Buffer |
| Physical Form | Liquid |
| Quantity | 25 mL |
For Research Use Only. Not for use in diagnostic procedures.