missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nuclear Fast Red, 94%, pure

CAS: 6409-77-4 | C14H11NNaO7S | 360.292 g/mol

$87.22 - $203.73

Chemical Identifiers

CAS 6409-77-4
Molecular Formula C14H11NNaO7S
Molecular Weight (g/mol) 360.292
MDL Number MFCD00078493
InChI Key KDEATZKCHKVXGA-UHFFFAOYSA-N
Synonym Calcium Red, C.I. 60760
PubChem CID 131675027
IUPAC Name 4-amino-1,3-dihydroxy-9,10-dioxoanthracene-2-sulfonic acid;molecular hydrogen;sodium
SMILES [HH].C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C(=C3N)O)S(=O)(=O)O)O.[Na]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC211980010
SDS View Documents
Thermo Scientific Chemicals
211980010
1 g Glass bottle
Each for $87.22
Only null left
 
AC211980050
SDS View Documents
Thermo Scientific Chemicals
211980050
5 g Glass bottle
Each for $203.73
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6409-77-4
360.292
KDEATZKCHKVXGA-UHFFFAOYSA-N
131675027
[HH].C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C(=C3N)O)S(=O)(=O)O)O.[Na]
C14H11NNaO7S
MFCD00078493
Calcium Red, C.I. 60760
4-amino-1,3-dihydroxy-9,10-dioxoanthracene-2-sulfonic acid;molecular hydrogen;sodium

Specifications

>300.0°C
92.0
94%
MFCD00078493
Solubility in water: 1g/L (25°C).
[HH].C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C(=C3N)O)S(=O)(=O)O)O.[Na]
360.292
357.26
Pure
1 g
6409-77-4
100.0
C14H11NNaO7S
Calcium Red, C.I. 60760
KDEATZKCHKVXGA-UHFFFAOYSA-N
4-amino-1,3-dihydroxy-9,10-dioxoanthracene-2-sulfonic acid;molecular hydrogen;sodium
131675027
94%
Glass bottle
Nuclear Fast Red
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 229-088-0

SDS
Documents

Documents

RUO – Research Use Only