missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nuclear Fast Red, 94%, pure

Catalog No. AC211980050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211980050 5 g Glass bottle
AC211980010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC211980050 Supplier Thermo Scientific Chemicals Supplier No. 211980050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6409-77-4 | C14H11NNaO7S | 360.292 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6409-77-4
Molecular Formula C14H11NNaO7S
Molecular Weight (g/mol) 360.292
MDL Number MFCD00078493
InChI Key KDEATZKCHKVXGA-UHFFFAOYSA-N
Synonym Calcium Red, C.I. 60760
PubChem CID 131675027
IUPAC Name 4-amino-1,3-dihydroxy-9,10-dioxoanthracene-2-sulfonic acid;molecular hydrogen;sodium
SMILES [HH].C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C(=C3N)O)S(=O)(=O)O)O.[Na]

Specifications

Melting Point >300.0°C
CAS Min % 92.0
CAS Max % 100.0
Packaging Glass bottle
Assay Percent Range 94%
Solubility Information Solubility in water: 1g/L (25°C).
Quantity 5 g
Formula Weight 357.26
Percent Purity 94%
Grade Pure
Chemical Name or Material Nuclear Fast Red
Show More Show Less
EINECSNumber 229-088-0

RUO – Research Use Only