Learn More
PeproTech NTF4 Polyclonal Antibody, Biotin, PeproTech®
Rabbit Polyclonal Antibody
$337.00 - $432.00
Specifications
| Antigen | NTF4 |
|---|---|
| Content And Storage | -20°C |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
50-252-9363
|
PeproTech
500P83BT25UG |
25 μg |
N/A
|
|
|||||
|
50-271-8444
|
PeproTech
500P83BT500UG |
500 μg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
50-252-9362
|
PeproTech
500P83BT50UG |
50 μg |
N/A
|
|
|||||
Description
AA Sequence of recombinant protein (monomer): GVSETAPASR RGELAVCDAV SGWVTDRRTA VDLRGREVEV LGEVPAAGGS PLRQYFFETR CKADNAEEGG PGAGGGGCRG VDRRHWVSEC KAKQSYVRAL TADAQGRVGW RWIRIDTACV CTLLSRTGRA Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human NT-4. Anti-Human NT-4-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human NT-4 by sandwich ELISA (using 100 μL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human NT-4 (500-P83) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human NT-4. Western Blot: To detect Human NT-4 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible development reagents the detection limit for Recombinant Human NT-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P83BT-1MG will be provided as 2 x 500 μg.
This gene is a member of a family of neurotrophic factors, neurotrophins, that control survival and differentiation of mammalian neurons. The expression of this gene is ubiquitous and less influenced by environmental signals. While knock-outs of other neurotrophins including nerve growth factor, brain-derived neurotrophic factor, and neurotrophin 3 prove lethal during early postnatal development, NTF5-deficient mice only show minor cellular deficits and develop normally to adulthood.Specifications
| NTF4 | |
| ELISA, Western Blot | |
| Biotin | |
| Rabbit | |
| Human | |
| NTF4 | |
| 2900040K06Rik; AI462899; GLC10; GLC1O; neurotrophic factor 4; neurotrophic factor 5; neurotrophin 4; Neurotrophin 4/5; neurotrophin 5; Neurotrophin 5 (neurotrophin 4/5); neurotrophin 5 (neurotrophin 4/5); neurotrophin-4; Neurotrophin-5; neutrophic factor 4; Neutrophic factor 5; NT4; NT-4; NT4/5; NT-4/5; NT4/NT5; NT4P; NT5; NT-5; Ntf4; Ntf5; Ntf-5 | |
| NTF4 | |
| Primary | |
| Affinity chromatography |
| -20°C | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| P34130 | |
| 4909 | |
| E.coli-derived, 28.1kDa Recombinant Human NT-4. | |
| Antibody |