missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTAN1 Antibody, ProSci
Description
NTAN1 Polyclonal Antibody specifically detects PDE10A in Human, Mouse, Rat Samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | NTAN1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | WB: 1:200 - 1:2000 |
| Formulation | PBS with 0.02% sodium azide, 50% glycerol, pH7.3. |
| Gene Alias | Protein N-terminal asparagine amidohydrolase, 351-, Protein NH2-terminal asparagine amidohydrolase, PNAA, Protein NH2-terminal asparagine deamidase, PNAD, Protein N-terminal Asn amidase, Protein N-terminal asparagine amidase, Protein NTN-amidase, NTAN1 |
| Host Species | Rabbit |
| Purification Method | Affinity purification |
| Quantity | 0.1 mL |
| Show More |