missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTAL Monoclonal Antibody (K52022_8E2), Invitrogen™
Mouse Monoclonal Antibody
$1,740.58 - $2,926.56
Specifications
| Antigen | NTAL |
|---|---|
| Clone | K52022_8E2 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: WGRFSKPPED DDANSYENVL ICKQKTTETG AQQEGIGGLC RGDLSLSLAL KTGPTSGLCP SASPEEDEES EDYQNSASIH QWRESRKVMG QLQREASPGP
NTAL Monoclonal antibody specifically detects NTAL in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| NTAL | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AW125574; HSPC046; LAB; Lat2; LAT-2 like; Linker for activation of B-cells; linker for activation of T cells family, member 2; linker for activation of T cells, transmembrane adaptor 2; linker for activation of T-cells family member 2; Membrane-associated adapter molecule; non-T cell activation linker; non-T-cell activation linker; NTAL; WBS15; WBSCR15; WBSCR5; Williams-Beuren syndrome chromosomal region 15 protein; williams-Beuren syndrome chromosomal region 15 protein homolog; Williams-Beuren syndrome chromosomal region 5 protein; Williams-Beuren syndrome chromosome region 5 homolog; Williams-Beuren syndrome chromosome region 5-like protein; WSCR5 | |
| Recombinant human LAT2 (immunogen range is 121-220). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| LAT2 |
| K52022_8E2 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9GZY6 | |
| 7462 | |
| IgG2b | |
| Protein A | |
| Antibody |