missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NSUN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa5115674
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115674 Supplier Invitrogen™ Supplier No. PA5115674
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NSUN2.

NSUN2 encodes a methyltransferase that catalyzes the intron-dependent formation of 5-methylcytosine at C34 of tRNA-leu(CAA). Maturation of cytoplasmic tRNAs includes splicing of introns, which are located 1 nucleotide 3-prime from the anticodon in all intron-containing tRNA genes. In tRNA-leu(CAA), the first position of the anticodon, C34, is converted to 5-methylcytosine, a modification necessary to stabilize the anticodon-codon pairing and correctly translate the mRNA. Mutations of NSUN2 result in autosomal recessive 5 mental retardation (MRT5).

Specifications

Antigen NSUN2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene NSUN2
Gene Accession No. Q08J23, Q1HFZ0
Gene Alias 5-methycytoisine methyltransferase; D13Wsu123e; hTrm4; Misu; mRNA cytosine C(5)-methyltransferase; MRT5; Myc-induced SUN domain containing protein; myc-induced SUN domain-containing protein; Myc-induced SUN-domain-containing protein; NOL1/NOP2/Sun domain family 2; NOL1/NOP2/Sun domain family member 2; NOL1/NOP2/Sun domain family, member 2; NOP2/Sun domain family, member 2; NOP2/Sun RNA methyltransferase family member 2; NOP2/Sun RNA methyltransferase family, member 2; Nsun2; RGD1311954; RNA cytosine C(5)-methyltransferase NSUN2; SAKI; substrate of AIM1/Aurora kinase B; TRM4; tRNA (cytosine(34)-C(5))-methyltransferase; tRNA (cytosine-5-)-methyltransferase NSUN2; tRNA cytosine C(5)-methyltransferase; tRNA methyltransferase 4 homolog
Gene Symbols NSUN2
Host Species Rabbit
Immunogen A synthesized peptide derived from human NSUN2(Accession Q08J23), corresponding to amino acid residues P504-E554.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28114, 361191, 54888
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less