missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NSMCE1 Monoclonal Antibody (K56018_3A9), Invitrogen™

Catalog No. MA556280
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556280 200 μL
MA555900 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556280 Supplier Invitrogen™ Supplier No. MA556280
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: MQGSTRRMGV MTDVHRRFLQ LLMTHGVLEE WDVKRLQTHC YKVHDRNATV DKLEDFINNI NSVLESLYIE IKRGVTEDDG RPIYALVNLA TTSISKMATD FAENELDLFR KALELIIDSE TGFASSTNIL NLVDQLKGKK MRKKEAEQVL QKFVQNKWLI EKEGEFTLHG RAILEMEQYI RETYPDAVKI CNICHSLLIQ GQSCETCGIR MHLPCVAKYF QSNAEPRCPH CNDYWPHEIP KVFDPEKERE SGVLKSNKKS LRSRQH

The NSMCE1 (Non-SMC Element 1 Homolog) gene encodes a protein that is a key component of the SMC5-SMC6 complex, which is essential for DNA repair and the maintenance of genomic stability. Located on chromosome 16, the NSMCE1 protein interacts with other components of the SMC5-SMC6 complex, including SMC5, SMC6, and NSMCE2, facilitating the repair of DNA double-strand breaks and promoting chromosome segregation during cell division. NSMCE1 is particularly involved in homologous recombination, a critical pathway for error-free DNA repair. Mutations or deficiencies in NSMCE1 can lead to genomic instability, increased sensitivity to DNA-damaging agents, and have been associated with several genetic disorders and cancer predispositions.

Specifications

Antigen NSMCE1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K56018_3A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene NSMCE1
Gene Accession No. Q8WV22
Gene Alias 2510027N19Rik; HSPC333; HSPC337; Non-SMC element 1 homolog; non-SMC element 1 homolog (S. cerevisiae); non-structural maintenance of chromosomes element 1 homolog; NSE1; NSE1 homolog, SMC5-SMC6 complex component; Nsmce1; RGD1307760
Gene Symbols NSMCE1
Host Species Mouse
Immunogen Recombinant human NSMCE1 (immunogen range is 1-266).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197370
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less