missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals NSC 663284
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 1 mg |
| Description | Red |
| Content And Storage | -30°C to -10°C |
| Formula Weight | 321.76g/mol |
| Sensitivity | Light Sensitive |
| Form | Crystalline solid |
| InChI Key | BMKPVDQDJQWBPD-UHFFFAOYSA-N |
| SMILES | ClC1=C(NCCN2CCOCC2)C(=O)C2=C(C=CC=N2)C1=O |
| IUPAC Name | 6-chloro-7-{[2-(morpholin-4-yl)ethyl]amino}-5,8-dihydroquinoline-5,8-dione |
| Molecular Weight (g/mol) | 321.76 |
Safety and Handling
| Recommended Storage | Store at -20°C |
RUO – Research Use Only