accessibility menu, dialog, popup

UserName

CAS RN 383907-43-5

IUPAC Name: 6-chloro-7-{[2-(morpholin-4-yl)ethyl]amino}-5,8-dihydroquinoline-5,8-dione
Synonyms: 6-chloro-7-{[2-(morpholin-4-yl)ethyl]amino}quinoline-5,8-dione
Molecular Weight (g/mol): 321.76
Molecular Formula: C15H16ClN3O3
InChi Key: BMKPVDQDJQWBPD-UHFFFAOYSA-N
SMILES: ClC1=C(NCCN2CCOCC2)C(=O)C2=C(C=CC=N2)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results