missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals NSC 663284

Catalog No. AAJ66520MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66520MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66520MCR Supplier Thermo Scientific Chemicals Supplier No. J66520MCR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 383907-43-5 | C15H16ClN3O3, C15H16ClN3O3 | 321.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 383907-43-5
Molecular Formula C15H16ClN3O3
Synonym 6-Chloro-7-(2-morpholin-4-yl-ethylamino)quinoline-5,8-dione; Cdc25 Phosphatase Inhibitor II

Specifications

Quantity 1 mg
Description Red
Content And Storage -30°C to -10°C
Formula Weight 321.76g/mol
Sensitivity Light Sensitive
Form Crystalline solid

RUO – Research Use Only