missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NS3, Mouse anti-Hepatitis C virus (HCV), Clone: 1B6, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16079
Specifications
| NS3 | |
| Monoclonal | |
| Unconjugated | |
| In PBS (10% glycerol, 0.02% sodium azide). | |
| Recombinant protein corresponding to serine protease domain of HCV NS3. | |
| RUO | |
| Recognizes HCV NS3 (genotype 1a and 1b) (epitope mapped to amino acid 160-193 of NS3), recognizes NS3 alone or the NS3-4A complex. Does not cross-react with HCV NS3 (genotype 2a). | |
| For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1B6 | |
| Immunocytochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Hepatitis C virus | |
| Antibody | |
| IgG1 |