missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NRSN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563920
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563920 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563920 Supplier Invitrogen™ Supplier No. PA563920
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: VKSYSKEEKF LQQKFKERIA DIKAHTQPVT KAPGPGETKI PVTLSRVQNV QPLLAT Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

May play an important role in neural organelle transport, and in transduction of nerve signals or in nerve growth. May play a role in neurite extension. May play a role in memory consolidation. [UniProt].

Specifications

Antigen NRSN1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NRSN1
Gene Accession No. Q8IZ57
Gene Alias neurensin 1; neurensin-1; Neuro-p24; nrsn1; p24; vesicular membrain protein p24; vesicular membrane protein of 24 kDa; Vesicular membrane protein p24; Vmp
Gene Symbols NRSN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NRSN1. Recombinant protein control fragment (Product #RP-101823).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140767
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less