missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nrcam Mouse anti-Human, Mouse, Rat, ATTO 594, Clone:S364-51, Abnova™
Mouse monoclonal antibody raised against partial recombinant mouse Nrcam.
Supplier: Abnova Corporation MAB17672
Specifications
| Nrcam | |
| Monoclonal | |
| Atto 594 | |
| Immunocytochemistry (1:100) Immunofluorescence (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| Nrcam | |
| Mouse | |
| 100 μg | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| S364-51 | |
| neuron-glia-CAM-related cell adhesion molecule | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| Bravo, C030017F07Rik, C130076O07Rik, mKIAA0343 | |
| Recombinant protein corresponding to amino acids 30-845 at extracellular domain of mouse Nrcam. | |
| 319504 | |
| Store at -20°C. | |
| Liquid |