missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NRBF2 Monoclonal Antibody (K40009_12G3), Invitrogen™

Catalog No. MA556174
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556174 200 μL
MA555794 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556174 Supplier Invitrogen™ Supplier No. MA556174
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: GQSPLSQKYS PSTEKCLPEI QGIFDRDPDT LLYLLQQKSE PAEPCIGSKA PKDDKTIIEE QATKIADLKR HVEFLVAENE RLRKENKQLK AEKARLLKGP IEKELDVDAD

NRBF2 (nuclear receptor-binding factor 2) modulates transcriptional activation by target nuclear receptors. It is involved in starvation-induced autophagy probably by its association with PI3K complex I (PI3KC3-C1). NRBF2 stabilizes the PI3KC3-C1 assembly and enhances ATG14-linked lipid kinase activity of PIK3C3.

Specifications

Antigen NRBF2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K40009_12G3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Nrbf2
Gene Accession No. Q96F24
Gene Alias Comodulator of PPAR and RXR; comodulator of PPAR and RXR 1; comodulator of PPAR and RXR 2; COPR; COPR1; COPR2; Nrbf2; NRBF-2; nuclear receptor binding factor 2; nuclear receptor binding factor-2; nuclear receptor-binding factor 2
Gene Symbols Nrbf2
Host Species Mouse
Immunogen Recombinant human NRBF2 (immunogen range is 111-220).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29982
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less