missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NR1D1 Recombinant Rabbit Monoclonal Antibody (9I2H1)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542757
Description
Positive test controls include: HepG2, Mouse liver, Rat brain, Rat lung. Immunogen sequence: SPEPTVEDVI SQVARAHREI FTYAHDKLGS SPGNFNANHA SGSPPATTPH RWENQGCPPA PNDNNTLAAQ RHNEALNGLR Q.
This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.Chemical Identifiers
| NR1D1 | |
| 1 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P20393, Q3UV55, Q63503 | |
| 217166, 252917, 9572 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 280-360 of human Rev-Erba/NR1D1 (NP_0683701). | |
| Antibody |
| 9I2H1 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| NR1D1 | |
| A530070C09Rik; BD73; Ear1; ear-1; EAR-1R; HREV; NR1D1; nuclear receptor protein; nuclear receptor Rev-ErbA-alpha; nuclear receptor subfamily 1 group D member 1; nuclear receptor subfamily 1, group D, member 1; orphan nuclear receptor; R75201; rev-erb alpha; Rev-erba; rev-erbA alpha; rev-erbA(alpha); Reverbaalpha; Rev-ErbAalpha; rev-erbA-alpha; Rev-ErbA-alpha protein; REV-ERBalpha; RVR; THRA1; THRAL; V-erbA-related protein 1; V-erbA-related protein EAR-1 | |
| NR1D1 | |
| Primary | |
| Affinity Chromatography |
Specifications
| NR1D1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P20393, Q3UV55, Q63503 | |
| NR1D1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 280-360 of human Rev-Erba/NR1D1 (NP_0683701). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 9I2H1 | |
| Unconjugated | |
| NR1D1 | |
| A530070C09Rik; BD73; Ear1; ear-1; EAR-1R; HREV; NR1D1; nuclear receptor protein; nuclear receptor Rev-ErbA-alpha; nuclear receptor subfamily 1 group D member 1; nuclear receptor subfamily 1, group D, member 1; orphan nuclear receptor; R75201; rev-erb alpha; Rev-erba; rev-erbA alpha; rev-erbA(alpha); Reverbaalpha; Rev-ErbAalpha; rev-erbA-alpha; Rev-ErbA-alpha protein; REV-ERBalpha; RVR; THRA1; THRAL; V-erbA-related protein 1; V-erbA-related protein EAR-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 217166, 252917, 9572 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |