missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NPTX1 Monoclonal Antibody (K92045_10E7), Invitrogen™

Catalog No. ma556013
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556013 200 μL
MA555633 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556013 Supplier Invitrogen™ Supplier No. MA556013
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: SQLGQTLQSL KTRLENLEQY SRLNSSSQTN SLKDLLQSKI DELERQVLSR VNTLEEGKGG PRNDTEERVK IETALTSLHQ RISELEKGQK

NPTX1 is a member of the neuronal pentraxin family. Human neuronal pentraxin 1 is similar to rat NP1 which is a binding protein for the snake venom toxin taipoxin.

Specifications

Antigen NPTX1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K92045_10E7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Nptx1
Gene Accession No. P47971, Q15818
Gene Alias 47 kDa taipoxin-binding protein; D11Bwg1004e; neuronal pentraxin 1; neuronal pentraxin I; Neuronal pentraxin-1; NP1; NP-I; NPTX1
Gene Symbols Nptx1
Host Species Mouse
Immunogen Recombinant human NPTX1 (immunogen range is 131-220).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 266777, 4884
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less