missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPTX1 Monoclonal Antibody (K92045_10E7), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555633
Description
Sequence of this protein is as follows: SQLGQTLQSL KTRLENLEQY SRLNSSSQTN SLKDLLQSKI DELERQVLSR VNTLEEGKGG PRNDTEERVK IETALTSLHQ RISELEKGQK
NPTX1 is a member of the neuronal pentraxin family. Human neuronal pentraxin 1 is similar to rat NP1 which is a binding protein for the snake venom toxin taipoxin.Specifications
| NPTX1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P47971, Q15818 | |
| Nptx1 | |
| Recombinant human NPTX1 (immunogen range is 131-220). | |
| 50 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| K92045_10E7 | |
| Unconjugated | |
| Nptx1 | |
| 47 kDa taipoxin-binding protein; D11Bwg1004e; neuronal pentraxin 1; neuronal pentraxin I; Neuronal pentraxin-1; NP1; NP-I; NPTX1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 266777, 4884 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |