missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOXO1 Polyclonal Antibody

Description
Recommended positive controls: HCT116. Predicted reactivity: Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NADPH oxidases catalyze the transfer of electrons from NADPH to molecular oxygen to generate reactive oxygen species to NOX and also target NOX to different subcellular compartments.
Specifications
Specifications
| Antigen | NOXO1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | NOXO1 |
| Gene Accession No. | Q8NFA2 |
| Gene Alias | 2310034C04Rik; hslt; MGC20258; NADPH oxidase organizer 1; NADPH oxidase regulatory protein; Nox organizer 1; NOXO1; Nox-organizing protein 1; P41NOX; P41NOXA; P41NOXB; P41NOXC; regulatory protein P41NOX; SH3 and PX domain-containing protein 5; SH3PXD5; Snx28; Sorting nexin-28 |
| Gene Symbols | NOXO1 |
| Show More |