missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOX2 Recombinant Rabbit Monoclonal Antibody (23GB5135)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA552800
Description
NOX2 Recombinant Monoclonal Antibody for Flow, Western Blot
Mox1 and the glycoprotein gp91-phox are largely related proteins that are essential components of the NADPH oxidase. The superoxide-generating NADPH oxidase complex expresses in phagocytes, neuroepithelial bodies, vascular smooth muscle cells, and endothelial cells. It includes a membrane bound flavocytochrome containing two subunits, gp91-phox and p22-phox, and the cytosolic proteins p47-phox and p67-phox. The p22- and gp91-phox subunits also function as surface O2 sensors that initiate cellular signaling in response to hypoxic conditions. Mox1 and gp91 contain identical C-terminal sequence identity, yet possess distinct expression patterns. gp91-phox expresses in eosinophils, neutrophils, monocytes, and B-lymphocytes, whereas Mox1 is predominantly detected in the colon, with low expression in the uterus and prostate.Chemical Identifiers
| NOX2 | |
| 1 mg/mL | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P04839, Q61093 | |
| 13058, 1536, 66021 | |
| A synthesized peptide derived from human NOX2/CYBB/gp91phox (142-204AA). | |
| Antibody |
| 23GB5135 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Cybb | |
| AMCBX2; C88302; Cgd; CGD91-phox; Cybb; Cyd; cytochrome b(558) subunit beta; cytochrome b-245 beta chain; cytochrome b-245 beta polypeptide; cytochrome b-245 heavy chain; cytochrome b-245, beta polypeptide; cytochrome b558 subunit beta; endothelial type gp91-phox; GP91 PHOX; GP911; GP91-1; GP91PHOX; gp91-phox; Heme-binding membrane glycoprotein gp91phox; IMD34; NADPH oxidase 2; neutrophil cytochrome b 91 kDa polypeptide; Nox2; p22 phagocyte B-cytochrome; p91PHOX; p91-PHOX; superoxide-generating NADPH oxidase heavy chain subunit | |
| Cybb | |
| Primary | |
| Affinity chromatography |
Specifications
| NOX2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P04839, Q61093 | |
| Cybb | |
| A synthesized peptide derived from human NOX2/CYBB/gp91phox (142-204AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot | |
| 23GB5135 | |
| Unconjugated | |
| Cybb | |
| AMCBX2; C88302; Cgd; CGD91-phox; Cybb; Cyd; cytochrome b(558) subunit beta; cytochrome b-245 beta chain; cytochrome b-245 beta polypeptide; cytochrome b-245 heavy chain; cytochrome b-245, beta polypeptide; cytochrome b558 subunit beta; endothelial type gp91-phox; GP91 PHOX; GP911; GP91-1; GP91PHOX; gp91-phox; Heme-binding membrane glycoprotein gp91phox; IMD34; NADPH oxidase 2; neutrophil cytochrome b 91 kDa polypeptide; Nox2; p22 phagocyte B-cytochrome; p91PHOX; p91-PHOX; superoxide-generating NADPH oxidase heavy chain subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13058, 1536, 66021 | |
| -20°C | |
| Liquid |