missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ WEHI-3 Whole Cell Lysate

Catalog No. NBP213176
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP213176 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP213176 Supplier Novus Biologicals™ Supplier No. NBP213176

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The WEHI-3 Whole Cell Lysate is derived from mouse. It has been validated for the following applications: SDS-Page.

Specifications

Tissue Blood Lymph
Quantity 0.25 mg
Cellular Fraction Whole
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native

For Research Use Only