missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ U-2 OS Whole Cell Lysate

Catalog No. NBP213171
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP213171 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP213171 Supplier Novus Biologicals™ Supplier No. NBP213171
The U-2 OS Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation, SDS-Page.

Specifications

Tissue Bone/Cartilage
For Use With (Application) Immunoprecipitation, SDS-Page, Western Blot
Quantity 0.25 mg
Storage Buffer HEPES (pH 7.9), KCl, Sucrose, Sodium deoxycholate, and NP-40 (without denaturing agents).
Purity Protein
Test Specificity U-2 OS (human osterogenic sarcoma) whole cell lysate (non-denatured).
Cellular Fraction Whole
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native

For Research Use Only