missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TLR2 Cell Lysate

Catalog No. NBP230165
Change view
Click to view available options
Quantity:
180 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP230165 180 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP230165 Supplier Novus Biologicals™ Supplier No. NBP230165

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The TLR2 Cell Lysate is derived from mouse. It has been validated for the following applications: Western Blot.

Specifications

Cell Line Mouse TLR2 Transfected Cell Lysate
For Use With (Application) Western Blot
Quantity 180 μL
Storage Buffer 1X Sample Buffer containing b-Mercaptoethanol, 1X SDS sample buffer: 60mM Tris-Hcl (pH 6.8), 10mM EDTA, 2 % SDS, 0.03% Bromophenol Blue, 10% glycerol, 0.3% b-mercaptoethanol
Host Species Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.

For Research Use Only